Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX1, Rabbit anti-Human,Mouse,Equine,Opossum,Primate,Xenopus, Biotin, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP224486B

Catalog No. NB005224


Add to cart

Description

Description

SOX1 Polyclonal specifically detects SOX1 in Human, Mouse, Equine, Opossum, Primate, Xenopus samples. It is validated for Western Blot.
Specifications

Specifications

SOX1
Biotin
PBS with 0.05% Sodium Azide
SRY (sex determining region Y)-box 1, SRY-related HMG-box gene 1, transcription factor SOX-1
Rabbit
IgG
0.1 mL
Stem Cells
Primary
Human, Mouse, Equine, Opossum, Primate, Xenopus
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Western Blot
Affinity Purified, Conjugated
SOX1
A portion of amino acids 100-150 of human SOX1 was used as the immunogen.
Immunogen affinity purified
RUO
Store at 4°C in the dark.
Polyclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit