Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX10 Mouse anti-Human, Mouse, PE/Cy5.5, Clone: SOX10/991, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP259621PECY55

Catalog No. N259621PEC5


Add to cart

Description

Description

SOX10 Monoclonal antibody specifically detects SOX10 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

SOX10
SOX10/991
Flow Cytometry
Conjugated, Purified
SOX10
Recombinant human SOX10 protein fragment (around aa115-269) (exact sequence is proprietar
Protein A or G purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
The specificity of this monoclonal antibody to human, mouse SOX10 was validated by HuProtTM Array, containing more than 19,000, full-length human proteins. Recognizes a protein of ∽55kDa, identified as SOX10. This MAb is highly specific and does not cross-react with other members of the SOX-family. SOX genes comprise a family of genes that are related to the mammalian sex-determining gene SRY. These genes similarly contain sequences that encode for the HMG -box domain, which is responsible for the sequence-specific DNA-binding activity. SOX-10 is a sensitive marker of melanoma, including conventional, spindled, and desmoplastic subtypes. It is expressed by metastatic melanomas and nodal capsular nevus in sentinel lymph nodes, but not by other lymph node components such as dendritic cells, which usually express S100 protein. Commonly used melanoma markers, such as anti-HMB-45 and anti-Melan-A, are poorly expressed in desmoplastic melanomas while SOX-10 is moderately to strongly expressed in desmoplastic melanomas. SOX-10 is considered as a very reliable marker for recognizing residual desmoplastic melanomas. In normal tissues, it is expressed in Schwann cells, melanocytes, and myoepithelial cells of salivary, bronchial and mammary glands. SOX-10 expression is also observed in mast cells
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
MGC15649, PCWH, SRY (sex determining region Y)-box 10, SRY-related HMG-box gene 10, transcription factor SOX-10, WS4C, WS4mouse, human homolog of
Mouse
IgG2b Kappa
0.1 mL
Primary
6663.0
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit