Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX10, Rabbit anti-Human,Mouse, DyLight 405, Clone: SOX10/2311R, Novus Biologicals
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP279905V
Description
SOX10 Monoclonal antibody specifically detects SOX10 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
SOX10 | |
SOX10/2311R | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Conjugated, Purified | |
SOX10 | |
A recombinant fragment (around aa115-269) of human SOX10 protein (exact sequence is proprietary) | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MGC15649, PCWH, SRY (sex determining region Y)-box 10, SRY-related HMG-box gene 10, transcription factor SOX-10, WS4C, WS4mouse, human homolog of | |
Rabbit | |
IgG | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title