Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX11 Rabbit anti-Human, Clone: SOX11/3235R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen SOX11
Clone SOX11/3235R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 2-4 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122850
View Documents
Novus Biologicals
NBP30768820UG
20 μg
Each for $159.82
Add to cart
 
NB122849
View Documents
Novus Biologicals
NBP30768810UG
100 μg
Each for $406.52
Add to cart
 
Description

Description

SOX11 Monoclonal antibody specifically detects SOX11 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

SOX11
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
10 mM PBS with 0.05% BSA
6664
IgG Kappa
Primary
Store at 4°C.
SOX11/3235R
Immunohistochemistry-Paraffin 2-4 ug/ml
Unconjugated
Rabbit
Human
SRY (sex determining region Y)-box 11, SRY (sex-determining region Y)-box 11, SRY-related HMG-box gene 11, transcription factor SOX-11
Recombinant fragment (around aa300-400) of human SOX11 protein
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit