Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SOX14 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA577173
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This intronless gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. Mutations in this gene are suggested to be responsible for the limb defects associated with blepharophimosis, ptosis, epicanthus inversus syndrome (BPES) and Mobius syndrome.
Specifications
SOX14 | |
Polyclonal | |
Unconjugated | |
SOX14 | |
HMG box transcription factor SOX-14; Protein SOX-28; SOX14; Sox-14; SOX28; SRY (sex determining region Y)-box 14; SRY-box 14; SRY-box containing gene 14; transcription factor SOX-14 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20669, 300954, 8403 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
O95416, Q04892 | |
SOX14 | |
KLH conjugated synthetic peptide derived from human SOX14: 51-150. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction