Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SOX17 Monoclonal Antibody (OTI2G8)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA524891
Description
SOX17 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
SOX17 (transcription factor SOX-17) belongs to the SRY-related HMB box family of transcription factors. It acts as a transcription regulator that binds target promoter DNA and bends the DNA. It binds to the sequences 5'-AACAAT-'3 or 5'-AACAAAG-3'. SOX17 modulates transcriptional regulation via WNT3A and inhibits Wnt signaling. It promotes degradation of activated CTNNB1. SOX17 plays an essential role in the regulation of embryonic development and is required for normal looping of the embryonic hear tube and for normal development of the definitive gut endoderm. Mutations in the gene can result in vesicoureteral reflux.
Specifications
SOX17 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9H6I2 | |
SOX17 | |
Human recombinant protein fragment corresponding to amino acids 177-414 of SOX17 produced in E.coli. | |
100 μL | |
Primary | |
Human, Canine | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
OTI2G8 | |
Unconjugated | |
SOX17 | |
HMG transcription factor SOX17; HMG-box transcription factor Sox17; Sox17; Sox-17; SRY (sex determining region Y)-box 17; SRY box 17; SRY-box 17; SRY-box containing gene 17; SRY-box transcription factor 17; SRY-related HMG-box transcription factor SOX17; transcription factor SOX-17; transcription factor SOX-17; LOW QUALITY PROTEIN: transcription factor SOX-17; VUR3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
486955, 64321 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction