Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX18 Rabbit anti-Human, Clone: SC05-85, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP267492
Description
SOX18 Monoclonal antibody specifically detects SOX18 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SOX18 | |
SC05-85 | |
Unconjugated | |
Purified | |
HLTS, SRY (sex determining region Y)-box 18, SRY-box 18, transcription factor SOX-18 | |
SOX18 | |
Recombinant protein | |
Monoclonal | |
Protein A purified | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1:1000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide | |
54345 | |
Rabbit | |
IgG | |
Primary | |
100μL | |
Human,Mouse,Rat |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title