Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

$206.50 - $455.38

Specifications

Antigen SOX2
Concentration 0.5 mg/mL
Dilution Western Blot 1-3 ug/ml, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 20 ug/ml
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224390SS
SDP
View Documents
Novus Biologicals
NBP224390SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP224390
SDP
View Documents
Novus Biologicals
NBP224390
0.1 mg
Each for $455.38
Only null left
Add to Cart
 
Description

Description

SOX2 Polyclonal specifically detects SOX2 in Human, Mouse, Porcine, Chicken, Feline, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

SOX2
Western Blot 1-3 ug/ml, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 20 ug/ml
Polyclonal
Rabbit
Cellular Markers, Core ESC Like Genes, Embryonic Stem Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
ANOP3, MCOPS3, MGC2413, SRY (sex determining region Y)-box 2, SRY-related HMG-box gene 2, transcription factor SOX2, transcription factor SOX-2
SOX2
IgG
Affinity Purified
The amino acid sequence used as immunogen for the Wnt5A antibody is 100% homologous in human, cow, monkey, rat and mouse, 94% in opossum, 88% in xenopus, 81% in chimpanzee and 76% homologous in pufferfish.
0.5 mg/mL
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse, Pig, Chicken, Feline, Primate
P48431
6657
A portion of amino acids 100-150 of human SOX2 was used as the immunogen for the antibody.
Primary
Store at -20C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.