Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX2 Mouse anti-Human, Alexa Fluor™ 350, Clone: 4G8, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP229623AF350
Description
SOX2 Monoclonal specifically detects SOX2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SOX2 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
SOX2 | |
Full length recombinant human SOX2 (NCBI NP_003097). | |
Primary | |
Human | |
IgG2b |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
4G8 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
ANOP3, MCOPS3, MGC2413, SRY (sex determining region Y)-box 2, SRY-related HMG-box gene 2, transcription factor SOX2, transcription factor SOX-2 | |
Mouse | |
0.1 mL | |
6657 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title