Learn More
SOX2 Mouse anti-Human, Alexa Fluor™ 750, Clone: 4G8, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP229623AF750
Catalog No. NP229623A70
Add to cart
Description
Description
SOX2 Monoclonal antibody specifically detects SOX2 in Human samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
SOX2
4G8
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
ANOP3, MCOPS3, MGC2413, SRY (sex determining region Y)-box 2, SRY-related HMG-box gene 2, transcription factor SOX2, transcription factor SOX-2
Mouse
IgG2b
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
SOX2
Full length recombinant human SOX2 (NCBI NP_003097).
0.1 mL
Primary
6657