Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX3 Mouse anti-Human, Clone: CL4725, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524710

Catalog No. PIMA524710


Add to cart

Description

Description

Immunogen sequence: RENSSGARSPR VPADLARSIL ISLPFPPDSL AHRPPSSAPT ESQGLFTVAA PAPGAPSPPA TLAHLLPAPA MYSLLETELK NPVGTPTQAA GTGGPAAPGG AGKSSA Binds to an epitope located within the peptide sequence LISLPFPPDS as determined by overlapping synthetic peptides. Highest antigen sequence identity to the following orthologs - mouse 82%, rat 31%.

This gene encodes a member of the SOX family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. Mutations in this gene have been associated with X-linked mental retardation with growth hormone deficiency.
Specifications

Specifications

SOX3
Monoclonal
1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P41225
SOX3; SRY (sex determining region Y)-box 3; GHDX; MRGH; PHP; PHPX; SOXB; transcription factor SOX-3
Mouse
IgG1
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
6658
Western Blot
CL4725
Unconjugated
SOX3
Liquid
SOX3
Recombit Human SOX3
Protein A
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only