Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP1507760.05MG
Description
SOX4 Polyclonal specifically detects SOX4 in Human samples. It is validated for Western Blot, ELISA.Specifications
SOX4 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.2), 0.5% BSA and 50% Glycerol with 0.03% Proclin | |
ecotropic viral integration site 16, EVI16, SRY (sex determining region Y)-box 4, SRY-related HMG-box gene 4, transcription factor SOX-4 | |
Rabbit | |
0.05 mg | |
6659 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
0.25 mg/mL | |
Western Blot, ELISA | |
Q06945 | |
SOX4 | |
Synthetic peptide selected from the N-terminal region of human SOX4 | |
Primary | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction