Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
$236.88 - $574.00
Specifications
Antigen | SOX4 |
---|---|
Concentration | 0.25 mg/mL |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP150776
|
Novus Biologicals
NBP150776 |
0.1 mg |
Each for $574.00
|
|
NBP15077605
|
Novus Biologicals
NBP1507760.05MG |
0.05 mg |
Each for $378.00
|
|
NB018670
|
Novus Biologicals
NBP1507760.025MG |
0.025 mg |
Each for $236.88
|
|
Description
SOX4 Polyclonal specifically detects SOX4 in Human samples. It is validated for Western Blot, ELISA.Specifications
SOX4 | |
Western Blot, ELISA | |
Unconjugated | |
Human | |
ecotropic viral integration site 16, EVI16, SRY (sex determining region Y)-box 4, SRY-related HMG-box gene 4, transcription factor SOX-4 | |
SOX4 | |
IgG |
0.25 mg/mL | |
Polyclonal | |
Rabbit | |
Q06945 | |
6659 | |
Synthetic peptide selected from the N-terminal region of human SOX4 | |
Primary |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title