Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SOX6 Monoclonal Antibody (CL5685)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA531425
Description
Immunogen sequence: NLDTFEHGGG HSYNHKQIE.
Transcriptional activator. Binds specifically to the DNA sequence 5'-AACAAT-3'. Plays a key role in several developmental processes, including neurogenesis and skeleton formation.
Specifications
SOX6 | |
Monoclonal | |
1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P35712 | |
SOX6 | |
Synthetic peptide corresponding to Human SOX6. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
CL5685 | |
Unconjugated | |
SOX6 | |
AI987981; HSSOX6; SOX6; Sox-6; SOXD; SOX-LZ; SRY (sex determining region Y)-box 6; SRY box 6; SRY-box 6; SRY-box containing gene 6; transcription factor SOX-6 | |
Mouse | |
Protein A | |
RUO | |
55553 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction