Learn More
Aviva Systems Biology Corp SOX8 Antibody - C-terminal region (P100732_P050) | 100uL
Supplier:  Aviva Systems Biology Corp P100732_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SOX8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SOX8. Alternate names: MGC24837ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-62695