Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX9, Mouse anti-Human, Clone: SOX9/2387, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP279769100UG

 View more versions of this product

Catalog No. NB010555


Add to cart

Description

Description

SOX9 Monoclonal antibody specifically detects SOX9 in Human Samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array.
Specifications

Specifications

SOX9
SOX9/2387
Unconjugated
10 mM PBS with 0.05% BSA
Purified
SOX9
Recombinant human SOX9 protein fragment (around aa 393-508) (exact sequence is proprietary)
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array, Western Blot
0.2 mg/mL
Western Blot 1-2 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
P48436, P48436
campomelic dysplasia, autosomal sex-reversal, CMD 1, CMD1, CMPD1, SRA1SRY (sex-determining region Y)-box 9 protein, SRY (sex determining region Y)-box 9, SRY-related HMG-box, gene 9, transcription factor SOX-9
Mouse
IgG1 Kappa
100 ug
Cancer, Phospho Specific, Stem Cell Markers
Primary
6662
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit