Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX9, Mouse anti-Human, Clone: SOX9/2387, Novus Biologicals™
Mouse Monoclonal Antibody
Specifications
Antigen | SOX9 |
---|---|
Clone | SOX9/2387 |
Concentration | 0.2 mg/mL |
Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array |
Description
SOX9 Monoclonal antibody specifically detects SOX9 in Human Samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array.Specifications
SOX9 | |
0.2 mg/mL | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
Unconjugated | |
Mouse | |
Cancer, Phospho Specific, Stem Cell Markers | |
10 mM PBS with 0.05% BSA | |
campomelic dysplasia, autosomal sex-reversal, CMD 1, CMD1, CMPD1, SRA1SRY (sex-determining region Y)-box 9 protein, SRY (sex determining region Y)-box 9, SRY-related HMG-box, gene 9, transcription factor SOX-9 | |
SOX9 | |
IgG1 κ | |
Protein A or G purified |
SOX9/2387 | |
Western Blot 1-2 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P48436, P48436 | |
6662 | |
Recombinant human SOX9 protein fragment (around aa 393-508) (exact sequence is proprietary) | |
Primary | |
Store at 4°C. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title