Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX9 Mouse anti-Human, Clone: SOX9/2398, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen SOX9
Clone SOX9/2398
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Applications Immunohistochemistry (Paraffin), Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122792
View Documents
Novus Biologicals
NBP30765920UG
20 μg
Each for $138.37
Add to cart
 
NB122791
View Documents
Novus Biologicals
NBP30765910UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

SOX9 Monoclonal antibody specifically detects SOX9 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array
Specifications

Specifications

SOX9
0.2 mg/ml
Immunohistochemistry (Paraffin), Peptide Array
Purified
RUO
Human
campomelic dysplasia, autosomal sex-reversal, CMD 1, CMD1, CMPD1, SRA1SRY (sex-determining region Y)-box 9 protein, SRY (sex determining region Y)-box 9, SRY-related HMG-box, gene 9, transcription factor SOX-9
Recombinant human SOX9 protein fragment (around aa 393-508) (exact sequence is proprietary)
Monoclonal
Protein A or G purified
SOX9/2398
Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Unconjugated
Mouse
Cancer, Phospho Specific, Stem Cell Markers
10 mM PBS with 0.05% BSA
6662
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit