Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX9 Rabbit anti-Human, Clone: SOX9/3916R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen SOX9
Clone SOX9/3916R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122794
View Documents
Novus Biologicals
NBP30766020UG
20 μg
Each for $159.82
Add to cart
 
NB122793
View Documents
Novus Biologicals
NBP30766010UG
100 μg
Each for $406.52
Add to cart
 
Description

Description

SOX9 Monoclonal antibody specifically detects SOX9 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

SOX9
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human
campomelic dysplasia, autosomal sex-reversal, CMD 1, CMD1, CMPD1, SRA1SRY (sex-determining region Y)-box 9 protein, SRY (sex determining region Y)-box 9, SRY-related HMG-box, gene 9, transcription factor SOX-9
Recombinant human SOX9 protein fragment (around aa 393-508) (exact sequence is proprietary)
Monoclonal
Protein A or G purified
SOX9/3916R
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Rabbit
Cancer, Phospho Specific, Stem Cell Markers
10 mM PBS with 0.05% BSA
6662
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit