Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ SP-D Antibody Pair [HRP]

Polyclonal/Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2795475PLATES

 View more versions of this product

Catalog No. NB010134


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes an antibody specific for Mouse SP-D.
Specifications

Specifications

62.5 pg/mL
Conjugated
6441
Antibody Pairs
62.5-4000 pg/mL
Storage is content dependent.
Rabbit
5 Plates
Sandwich ELISA
COLEC7collectin-7, collectin 7, Collectin-7, Lung surfactant protein D, PSPD, PSP-D, SFTP4pulmonary surfactant-associated protein D, SP-Dpulmonary surfactant apoprotein, surfactant protein D, surfactant, pulmonary-associated protein D, surfactant-associated protein, pulmonary 4
HRP
Sandwich ELISA
  • Detection Antibody: 0.2 mg/mL of rabbit anti-SP-D polyclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Capture Antibody: 1 mg/mL of rabbit anti-Mouse SP-D monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 105 ng of recombinant Mouse SP-D.
  • RUO
    This solid phase sandwich ELISA utilizes an antibody specific for Mouse SP-D.
    SP-D
    Cell Cycle and Replication
    Mouse
    SFTPD
    1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.6 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 4000 pg/mL is recommended.
    SDS
    Documents
    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit