Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SP-D Mouse anti-Human, Alexa Fluor™ 350, Clone: 10B2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223521AF350

Catalog No. NP22321A30


Add to cart

Description

Description

SP-D Monoclonal antibody specifically detects SP-D in Human samples. It is validated for Western Blot,ELISA.
Specifications

Specifications

SP-D
10B2
Western Blot, ELISA
COLEC7collectin-7, collectin 7, Collectin-7, Lung surfactant protein D, PSPD, PSP-D, SFTP4pulmonary surfactant-associated protein D, SP-Dpulmonary surfactant apoprotein, surfactant protein D, surfactant, pulmonary-associated protein D, surfactant-associated protein, pulmonary 4
Mouse
IgG2a Kappa
Store at 4°C in the dark.
Monoclonal
Clone 10B2 is specific for SP-D from amniotic fluid, serum and bronchioalveolar lavage. Clone 10B2 does not influence SP-D′s binding to carbohydrates, but block binding to GP-340.
ELISA, Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
SFTPD
Native human SP-D purified from late pregnancy amniotic fluid adsorbed onto aluminum hydroxide gel
0.1 mL
Primary
6441
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit