Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SP-D Mouse anti-Human, Janelia Fluor 549, Clone: 12G5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223563JF549

Catalog No. NBP23563549


Add to cart

Description

Description

SP-D Monoclonal antibody specifically detects SP-D in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

SP-D
12G5
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
SFTPD
Recombinant neck CRD SP-D adsorbed onto aluminum hydroxide
Protein A or G purified
Cell Cycle and Replication
Primary
6441
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
COLEC7collectin-7, collectin 7, Collectin-7, Lung surfactant protein D, PSPD, PSP-D, SFTP4pulmonary surfactant-associated protein D, SP-Dpulmonary surfactant apoprotein, surfactant protein D, surfactant, pulmonary-associated protein D, surfactant-associated protein, pulmonary 4
Mouse
IgG1 Kappa
0.1mL
Store at 4°C in the dark.
Monoclonal
Clone 12G5 is specific for human SP-D
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit