Learn More
Wilmad™ SP Scienceware™ 3mm O.D. Thin Walled Precision NMR Tube Available on GSA/VA Contract for Federal Government customers only.
To maximize SNR, Precision NMR Tubes have minimal paramagnetic impurities that would impact shimming. Tight I.D. and O.D. tolerance as small as 0.0065mm accommodates Wilmad inserts
Manufacturer:  Wilmad™ 663000385
 View more versions of this product
Catalog No. 16-800-293
  Check Availability
Add to cart
Alert:
Caps are not recommended for use with samples dissolved in chloroform-d or acetone-d6.
Specifications
Description
To maximize SNR, Precision NMR Tubes have minimal paramagnetic impurities that would impact shimming. Tight I.D. and O.D. tolerance as small as 0.0065mm accommodates Wilmad inserts
Description & Specifications
Specifications
203.2mm
2.9935±0.0065mm
2.4195±0.0065mm
8 in.
307-PP-8-5
51µm
25µm
300MHz
0.29mm