Learn More
Wilmad™ SP Scienceware™ 4mm O.D. Thin Walled Precision NMR Tubes Available on GSA/VA Contract for Federal Government customers only.
To maximize SNR, Precision NMR Tubes have minimal paramagnetic impurities that would impact shimming. Tight I.D. and O.D. tolerance as small as 0.0065mm accommodates Wilmad inserts
Manufacturer:  Wilmad™ 664000495
 View more versions of this product
Catalog No. 16-800-380
  Check Availability
Add to cart
Alert:
Caps are not recommended for use with samples dissolved in chloroform-d or acetone-d6.
Specifications Description & Specifications
Specifications
Borosilicate glass
3.9835±0.0065mm
25μm
400MHz
0.38mm
228.6mm
25μm
3.240±0.013mm
9 in.
427-PP-9-5