Learn More
Wilmad™ SP Scienceware™ 5mm O.D. Medium Walled Precision NMR Tube Available on GSA/VA Contract for Federal Government customers only.
To maximize SNR, Precision NMR Tubes have minimal paramagnetic impurities that would impact shimming. Tight I.D. and O.D. tolerance as small as 0.0065mm accommodates Wilmad inserts
Manufacturer:  Wilmad™ 661020595
 View more versions of this product
Catalog No. 16-800-277
  Check Availability
Add to cart
Alert:
Caps are not recommended for use with samples dissolved in chloroform-d or acetone-d6.
Specifications Description & Specifications
Specifications
Borosilicate glass
76μm
51μm
100MHz
0.77mm
228.6mm
4.9635±0.0067mm
3.43±0.15mm
9 in.
503-PS-9-5