Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bel-Art™ Clavies™ Biohazard Bag Holders

Click to view available options
Height (English):
18.5 in.
21.5 in.
27.2 in.
Size:
Small
Medium
Large
Description
- Economical, lightweight stainless-steel wire frame
- Highly visible orange polypropylene bottom tray for easy identification as a biohazard containment system
- Tray serves as containment well in case of bag leakage
- Autoclavable at 121°C (250°F) and dishwasher safe
- Assembly is easy and permanent
- Medium and large models can be used with autoclavable polypropylene cover(Mfr. No. F13192-0102) to improve sanitation
Specifications
Specifications
Autoclavable | Autoclavable |
Color | Silver, Orange |
Disposable | Reusable |
For Use With (Application) | Designed to securely support the bag and its contents |
For Use With (Equipment) | 01-826A, 01-826-5, 01-827B, 01-814-1, 01-815A small 30 x 61 cm Clavies biohazard bag |
Includes | Stainless Steel wire frame and orange Polypropylene bottom, plastic wire anchors |
Material | Stainless Steel |
Sterility | Sterile |
Size | Small |
Width (English) | 14 in. |
Show More |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction