Learn More
Bel-Art™ SP Scienceware™ Disposable Pipetting Reservoirs
Disposable and economical plastic pipetting reservoir. The SP Scienceware™ Disposable Pipetting Reservoirs includes ample clearance for all types of pipettors.
Specifications
Dimensions (L x W x H) 6 x 3.62 x 1.25 in. (15.2 x 9.2 x 3.2cm)
Capacity (Metric) 100 mL
Disposable Yes
Type Pipetting Reservoir
View More Specs
Products 2
Catalog Number Mfr. No. Sterility Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Sterility Packaging Price Quantity & Availability  
S97834
missing translation for 'viewDocuments'
Bel-Art™
F378770000/EMD
Non-sterile Bulk This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
S43310 Products for Science Education
missing translation for 'viewDocuments'
Bel-Art™
F378770001/EMD
Sterile Individually Wrapped This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • Ample clearance for all types of pipettors
  • Used for extended multichannel pipetting into microwell plates, Mini-Tubes, Cube 2ube™ Tubes and deep well blocks
  • 100ml capacity
Specifications
Specifications
6 x 3.62 x 1.25 in. (15.2 x 9.2 x 3.2cm)
100 mL
Yes
Pipetting Reservoir