Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Gas Purge Secador™ Amber Desiccator Cabinet Packages

Bundled units provide a cost savings over purchasing multiple units separately and provide greater space for dry storage needs

Specifications

Color Amber
Depth (Metric) 41.6 cm
Material Copolyester
Door Style Left-swinging
For Use With (Application) Provide a cost savings over purchasing multiple units separately and provide greater space for dry storage needs
View More Specs

Products 1
Catalog Number Mfr. No. Height (English) Height (Metric) Includes Volume (English) Price Quantity & Availability  
Catalog Number Mfr. No. Height (English) Height (Metric) Includes Volume (English) Price Quantity & Availability  
08-647-725
View Documents
Bel-Art™
F420740440
40.7 in. 103.6 cm Two 4.0 cabinet with rear mounted gas ports, latches and security loops for a padlock or tamper-evident seals, door-mounted digital hygrometer 3.74 cu. ft. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description

Description

Assures easy and reliable storage of moisture and UV sensitive products.

  • Durastar™ co-polyester construction blocks 99% of UV light and is resistant to staining, crazing and chemical attack
  • Rear mounted gas ports accept tubing with 1/4 to 3/8 in. I.D.
  • Amber color reduces visible light penetration by over 50% making it a good choice for storing light sensitive materials like reagents, analytical standards and investigational compounds
  • Large doors maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
  • Perforated shelves facilitate circulation of dry air; space for desiccant under bottom shelf
  • Easy-to-view digital hygrometer is located in the door front; side swinging door opens to the left
  • Compact 34.1W x 41.4cmD footprint for space saving stacking up to 3 units high
  • Gas ports can be closed to use desiccator with standard desiccant cartridges, F420450250, F420450200, F420480100
  • U.S. Patents: 6,834,920; 7,318,630 and D474,626

Specifications

Specifications

Amber
Copolyester
Provide a cost savings over purchasing multiple units separately and provide greater space for dry storage needs
41.6 cm
Left-swinging
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit