Bel-Art™ SP Scienceware™ Magnetic Bead Separation Racks

Powerful racks sequester magnetic beads so that affinity ligands or rinse solutions can be added or decanted in assays.

Manufacturer: Bel-Art™ F199000003

 View more versions of this product

Catalog No. 13-717-265

  Check Availability
Add to Cart

Alert

These separation racks contain powerful Neodymium magnets. Misuse can result in injury.

  • Do not use this product if you have a pacemaker or any other medical magnetized implant
  • Keep away from tools and objects that could be damaged by the magnetic field
  • Racks should be handled and stored away from devices subject to magnetic damage including credit cards and other products containing magnetic recording devices
  • Keep away from delicate instruments, watches, electronic equipment, displays and monitors
  • Magnets may attract steel or other magnetic material with high mechanical forces
  • Avoid contact between two magnets
  • Do not pull the magnets apart if contact has been made; twist off to prevent damage to the unit or fingers


    Specifications

    For Use With (Application) General Laboratories, Immunology, Proteomics, Genomics, Separation Methods
    Holds One 96-well PCR tube plate
    Material Neodymium magnets, Acrylic
    Product Type Magnetic Bead Separation Rack
    Color Clear
    Positions 1

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit