Learn More
Bel-Art™ SP Scienceware™ Micro-Mill™ Grinder Blades
Replacement blades for MicroMill grinder
Supplier:  Bel-Art™ H372580000
 View more versions of this product
Catalog No. 08-335-4
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
Stainless-steel or hard-faced blades.
Certifications
CE Marked
Specifications
Specifications
Replacement Blade
MicroMill grinder
12,000 rpm
Ideal for processing minerals, pharmaceuticals, pigment blends and other solid samples
Hard Faced
Product Suggestions
Product Suggestions