Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Octagon Spinbar™ Magnetic Stirring Bars
SCI_ED

Octagon magnetic stirring bars with integral pivot ring are the most commonly used shape for stirring small volumes in vessels such as vials, tubes and gradient makers.

 View more versions of this product

Catalog No. S68072



Add to cart

Description

Description

Used in conjunction with a magnetic stirring plate, Spinbar™ stir bars enable uniform, consistent, and thorough mixing of chemical solutions.
  • Multi-faceted shape provides greater surface area and added turbulence for more vigorous, thorough mixing when compared to the smooth surface of cylindrical bars
  • Pivot ring allows the Spin Bar to “rock” while spinning for better mixing conditions
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon™ brand PTFE coating; Superior chemical compatibility
  • Autoclavable
  • Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
Specifications

Specifications

50
0.31 in.
8mm
Yes
0.5 in.
Alnico Magnet
USP Class VI, ISO 9001:2008, SGS
White
Integral pivot ring
12.7mm
Octagonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit