Learn More
Fisherbrand™ Octagon Spinbar™ Magnetic Stirring Bars
GSA_VASCI_ED
Octagon magnetic stirring bars with integral pivot ring are the most commonly used shape for stirring small volumes in vessels such as vials, tubes and gradient makers.
$6.06 - $13.13
Specifications
Autoclavable Yes
Quantity 50
Certifications/Compliance USP Class VI, ISO 9001:2008, SGS
Color White
Material Alnico Magnet
View More Specs
Products 5
Catalog Number Diameter (English) Diameter (Metric) Length (English) Length (Metric) Price Quantity & Availability  
Catalog Number Diameter (English) Diameter (Metric) Length (English) Length (Metric) Price Quantity & Availability  
S68072
SCI_ED
View Documents
0.31 in. 8mm 0.5 in. 12.7mm
Each for $6.06
Case of 50 EA for $269.85
Add to cart
S68073
SCI_ED
View Documents
0.31 in. 0.8cm 0.63 in. 1.6mm
Each for $6.20
Case of 50 EA for $277.39
Add to cart
S68070
SCI_ED
View Documents
0.31 in. 8mm 1.5 in. 38mm
Each for $7.42
Case of 50 EA for $332.07
Add to cart
S68071
SCI_ED
View Documents
0.38 in. 9.5mm 2 in. 50.8mm
Each for $13.13
Case of 50 EA for $584.43
Add to cart
S68069
GSA_VASCI_ED
View Documents
0.31 in. 8mm 1 in. 25.4mm
Each for $6.45
Case of 50 EA for $287.50
Add to cart
Description
Description
Used in conjunction with a magnetic stirring plate, Spinbar™ stir bars enable uniform, consistent, and thorough mixing of chemical solutions.
  • Multi-faceted shape provides greater surface area and added turbulence for more vigorous, thorough mixing when compared to the smooth surface of cylindrical bars
  • Pivot ring allows the Spin Bar to “rock” while spinning for better mixing conditions
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon™ brand PTFE coating; Superior chemical compatibility
  • Autoclavable
  • Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
Specifications
Specifications
Yes
USP Class VI, ISO 9001:2008, SGS
Alnico Magnet
50
White
Octagonal