Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Plastic Tubing Cutter: Replacement Blades Products for Science Education

Hardened steel replacement blade for use with the Bel-Art™ Pocket-Sized Tubing Cutter S43338.

Manufacturer:  Bel-Art™ H210110000/EMD

Catalog No. S43339


Add to cart

Description

Description

  • Hardened steel blade makes clean, straight cuts
  • Blades measure: 76mm long, 24 per case
Specifications

Specifications

Steel
76.2 mm
Tubing cutter
3 in.
Tubing Cutter Replacement Blades
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit