Learn More
Bel-Art™ SP Scienceware™ Polyethylene Utility Bags Products for Science Education
Manufacturer:  Bel-Art™ H131780612/EMD
 View more versions of this product
Catalog No. S31800B
Add to cart
Description
Description
  • FDA approved material; Latex free
  • Different sizes available to best match particular packaging requirements
  • Not autoclavable
Order Info
Packed 100 per box
Specifications
Specifications
No
12 in.
PE (Polyethylene)
6 in.
4 mil
12 x 6 in. (305 x 152mm)
Shipping, storage and sampling
305mm
0.1016mm
152mm
Clear