Learn More
Bel-Art™ SP Scienceware™ Polyethylene Utility Bags
Reusable, transparent bags are well-suited for shipping or storage and are especially useful as sample bags
Supplier: Bel-Art™ H131761014/EMD
Description
- FDA approved material; Latex free
- Different sizes available to best match particular packaging requirements
- Not autoclavable
Specifications
Polyethylene | |
14 x 10 in. (356 x 254mm) | |
356 mm | |
254 mm |
0.038 mm | |
14 in. | |
10 in. |