Learn More
Bel-Art™ SP Scienceware™ Polyethylene Utility Bags
SCI_ED
Reusable, transparent bags are well-suited for shipping or storage and are especially useful as sample bags
$29.75 - $94.00
Specifications
Material Polyethylene
View More Specs
Products 20
Catalog Number Mfr. No. Dimensions (L x W) Thickness (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Thickness (Metric) Price Quantity & Availability  
S42881
SCI_ED
View Documents
Bel-Art™
H131760812/EMD
12 x 8 in. (305 x 203mm) 0.038 mm
Pack of 100 for $37.00
Add to Cart
S42883
SCI_ED
View Documents
Bel-Art™
H131760818/EMD
18 x 8 in. (457 x 203mm) 0.038 mm
Pack of 100 for $38.50
Add to Cart
S42884
SCI_ED
View Documents
Bel-Art™
H131761012/EMD
12 x 10 in. 0.038 mm
Pack of 100 for $34.50
Add to Cart
S42887
SCI_ED
View Documents
Bel-Art™
H131761218/EMD
18 x 12 in. (457 x 305mm) 0.038 mm
Pack of 100 for $47.00
Add to Cart
S42886
SCI_ED
View Documents
Bel-Art™
H131761016/EMD
16 x 10 in. (406 x 254mm) 0.038 mm
Pack of 100 for $43.25
Add to Cart
S42885
SCI_ED
View Documents
Bel-Art™
H131761014/EMD
14 x 10 in. (356 x 254mm) 0.038 mm
Pack of 100 for $36.50
Add to Cart
S42889
SCI_ED
View Documents
Bel-Art™
H131780046/EMD
4 x 6 in. (102 x 152mm) 0.102 mm
Pack of 100 for $29.75
Add to Cart
S42891
SCI_ED
View Documents
Bel-Art™
H131780610/EMD
6 x 10 in. (152 x 254mm) 0.102 mm
Pack of 100 for $45.75
Add to Cart
S42893
SCI_ED
View Documents
Bel-Art™
H131780812/EMD
8 x 12 in. (203 x 305mm) 0.102 mm
Pack of 100 for $51.25
Add to Cart
S42894
SCI_ED
View Documents
Bel-Art™
H131780816/EMD
16 x 8 in. (406 x 203mm) 0.102 mm
Pack of 100 for $66.75
Add to Cart
S42895
SCI_ED
View Documents
Bel-Art™
H131781012
10 x 12 in. (254 x 305mm) 0.102 mm
Pack of 100 for $60.50
Add to Cart
S42896
SCI_ED
View Documents
Bel-Art™
H131781016/EMD
10 x 16 in. (254 x 406mm) 0.102 mm
Pack of 100 for $81.00
Add to Cart
S42897
SCI_ED
View Documents
Bel-Art™
H131781018/EMD
18 x 10 in. (457 x 254mm) 0.102 mm
Pack of 100 for $82.00
Add to Cart
S42901
SCI_ED
View Documents
Bel-Art™
H131781218/EMD
18 x 12 in. (457 x 305mm) 0.102 mm
Pack of 100 for $88.00
Add to Cart
S42902
SCI_ED
View Documents
Bel-Art™
H131781220/EMD
12 x 20 in. (305 x 508mm) 0.102 mm
Pack of 100 for $94.00
Add to Cart
S42880
SCI_ED
View Documents
Bel-Art™
H131760612/EMD
12 x 6 in. (305 x 152mm) 0.038 mm Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View alternate product
N/A
S42882
SCI_ED
View Documents
Bel-Art™
H131760816/EMD
16 x 8 in. (406 x 203mm) 0.038 mm Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View alternate product
N/A
S42888
SCI_ED
View Documents
Bel-Art™
H131761220/EMD
20 x 12 in. (508 x 305mm) 0.038 mm Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View alternate product
N/A
S42892
SCI_ED
View Documents
Bel-Art™
H131780810/EMD
10.5 x 8 in. (267 x 203mm) 0.102 mm Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View alternate product
N/A
S42890
SCI_ED
View Documents
Bel-Art™
H131780068/EMD
8.5 x 6 in. (216 x 152mm) 0.102 mm Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View alternate product
N/A
Description
Description
  • FDA approved material; Latex free
  • Different sizes available to best match particular packaging requirements
  • Not autoclavable
Specifications
Specifications
Polyethylene