Learn More
Wilmad-LabGlass™ SP Scienceware™ PTFE NMR Tube Caps
Machined to exact specifications for greater spinning stability Wilmad-LabGlass PTFE NMR Tube Caps have a smaller gyroradius, more homogeneous mass distribution, and better chemical resistance than disposable caps.
Manufacturer:  Wilmad-LabGlass™ 660030025
 View more versions of this product
Catalog No. 16-800-213
Add to cart
Alert:
Fits only Wilmad-LabGlass Precision tubes due to the tight tolerance.
Description
Description
Recommended for experiments at high to ultra-high field and those using Chloroform as the reference solution.
  • Machined to exact specifications for greater spinning stability
  • Smaller gyroradius, more homogeneous mass distribution, and better chemical resistance than disposable caps
  • Fits tube sizes ranging from 3.0mm to 12.0mm
  • Fits Wilmad-LabGlass Precision Brand NMR Tubes only due to the tight tolerance
Specifications
Specifications
Not Autoclavable
White
Experiments at high to ultra-high field and those using Chloroform as the reference solution.
3 mm Wilmad NMR Tubes
PTFE
3.0 mm
25/Pk.