Learn More
Bel Art™ Round Lead Ring Flask Weights with Vikem™ Vinyl Coating
SCI_ED
Protects and stabilizes expensive glassware. Round Lead Ring Flask Weights with Vikem™ Vinyl Coating reduces floating, breakage and chemical spills.
$66.25 - $83.74
Specifications
Product Type Lead Ring Flask Weight
Autoclavable Not Autoclavable
Color Red
Material Vinyl-coated Lead
Shape Ring
View More Specs
Products 2
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Weight (English) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Weight (English) Price Quantity & Availability  
S31719B
SCI_ED
View Documents
Bel Art™
F183070010/EMD
2 in. 3.5 in. 1 lb.
Each for $83.74
Only null left
Add to Cart
S31719A
SCI_ED
View Documents
Bel Art™
F183070005
1.69 in. 2.9 in. 0.15 lb.
Each for $66.25
Only null left
Add to Cart
Description
Description
  • Stabilize glassware immersed in water baths; reduces floating, breakage and chemical spills
  • Bright red Vikem vinyl coating
  • Chemical-resistant
  • Will not scratch flasks
Specifications
Specifications
Lead Ring Flask Weight
Red
Ring
Yes
Erlenmeyer-style straight-necked flasks
Not Autoclavable
Vinyl-coated Lead
None
Non-sterile
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.