Learn More
Bel-Art™ SP Scienceware™ Secador™ Standard Desiccator Cabinets: Horizontal Models
A UV light-blocking, dry environment
$1484.71 - $1525.66
Specifications
Depth (English) | 16.3 in. |
---|---|
Depth (Metric) | 41.4 cm |
Autoclavable | Not Autoclavable |
Material | Copolyester |
No. of Shelves | 2 |
Catalog Number | Mfr. No. | Color | Shelf Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Shelf Type | Price | Quantity & Availability | ||||
08-647-58
|
Bel-Art™
F420740000 |
Clear | Adjustable Perforated |
Each for $1,484.71
|
|
08-647-669
|
Bel-Art™
F420740007 |
Blue with Clear Door | Vented Adjustable |
Each for $1,525.66
|
|
Description
The attractive design and sturdy construction of the Secador line of desiccator cabinets assures easy and reliable storage of moisture and light sensitive products.
- Durastar™ co-polyester construction blocks 99% of UV light and resists staining, crazing and chemical attack
- Amber color models reduce visible light penetration by over 50%, making them outstanding choices for storing light sensitive materials like reagents, analytical standards and investigational compounds
- Large doors maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
- Perforated shelves facilitate circulation of dry air; Space for desiccant under bottom shelf
- Easy-to-view hygrometer is located in the door front
- 4.0 horizontal has a wider footprint, lower height and pull down “oven style” door
- U.S. Patents: 6,834,920; 7,318,630 and D474,626
Dimensions
- Secador 1.0: 0.74 cu. ft. internal volume, 16.3L x 13.4W x 8.4 in.H
- Secador 2.0: 1.16 cu. ft. internal volume, 16.3L x 13.4W x 12.4 in.H
- Secador 3.0: 1.55 cu. ft. internal volume, 16.3L x 13.4W x 16.4 in.H
- Secador 4.0: 1.87 cu. ft. internal volume, 16.3L x 13.4W x 20.4 in.H
Order Info
Replacement shelves are available: Replacement Shelf for Horizontal 4.0 Cabinets, Mfr. No. 420740001 or Replacement Shelf for Vertical 1.0, 2.0, 3.0 or 4.0 Cabinets, Mfr. No. 420741001. Use with desiccant cartridges Mfr. Nos. 420450250, 420450200, 420480100.
Specifications
16.3 in. | |
Not Autoclavable | |
2 | |
Chemicals, stains and fine cracks |
41.4 cm | |
Copolyester | |
Easy and reliable storage of moisture and light sensitive products, blocks 99% UV light, used with disposable or reusable desiccant 420450250 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.