Learn More
Bel-Art™ SP Scienceware™ Transpette™ Eight Channel Transfer Pipettor
Perform plate washing, media change and supernatant collection without the use of pipettors and pipette tips with this inexpensive, eight-channel transfer pipettor
$70.50 - $228.00
Specifications
Volume (Metric) 600 μL
Disposable Yes
View More Specs
Products 2
Catalog Number Mfr. No. Sterility Material Quantity Packaging Type Price Quantity & Availability  
Catalog Number Mfr. No. Sterility Material Quantity Packaging Type Price Quantity & Availability  
S42970 Products for Science Education
View Documents
Bel-Art™
F378730001/EMD
Sterile Polyethylene 100/Cs. Individually Wrapped Case of 100 for $228.00
Add to cart
S32146 Products for Science Education
View Documents
Bel-Art™
F378730000/EMD
Non-sterile Polypropylene 25/Pk. Bulk Pack of 25 for $70.50
Add to cart
Description
Description
  • 8-Channel with 600µL capacity per channel
  • Made of translucent plastic
  • Discard after “hot” radioactive uses to avoid radiation contamination of expensive liquid handling devices
  • Non-autoclavable
  • Available with sterile and non-sterile options
Specifications
Specifications
600 μL
Yes