Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SP140L Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | SP140L |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP180140
|
Novus Biologicals
NBP180140 |
100 μL |
Each for $436.00
|
|
NBP18014020
|
Novus Biologicals
NBP18014020UL |
20 μL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
SP140L Polyclonal specifically detects SP140L in Human samples. It is validated for Western Blot.Specifications
SP140L | |
Polyclonal | |
Purified | |
RUO | |
NP_612411 | |
93349 | |
Synthetic peptide directed towards the middle region of human LOC93349. Peptide sequence TVDFQAPLLPVTCGGVKGILHKEKLEQGTLAKCIQTEDGKWFTPMEFEIK. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
MGC132667, nuclear body protein SP140-like protein, SP140 nuclear body protein-like | |
SP140L | |
IgG | |
Protein A purified | |
39 kDa |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title