Learn More
Cygnus TechnologiesSupplier Diversity Partner SP2/0 HCP ELISA KIT DFS Item
Manufacturer:  Cygnus TechnologiesSupplier Diversity Partner F180
Recombinant expression by the hybridoma cell line SP2/0 is a relatively simple and cost effective method for production of monoclonal antibodies intended for use as therapeutic agents in humans. Our assay validation has determined that the LOD is 0.1ng/mL.
Catalog No. NC0199194