Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SP6 RNA Polymerase in Potassium Phosphate Buffer Kit

Transcribes RNA from DNA sequences

Specifications

CAS 9014-24-8
Synonym RNA Polymerase from Salmonella enterica serotype typhimurium, SP6-infected
Storage Requirements −30° to −10°C
Format Liquid
For Use With (Application) Transcribes RNA from DNA sequences
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ65573OAT View Documents Alfa Aesar™
J65573OAT
5 Kilounits This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Specifications

Specifications

9014-24-8
−30° to −10°C
Transcribes RNA from DNA sequences
RNA Polymerase
RNA Polymerase from Salmonella enterica serotype typhimurium, SP6-infected
Liquid
Colorless
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit