Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPACA5 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548153

Catalog No. PA548153

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Hydrolysis of (1->4)-beta-linkages between N-acetylmuramic acid and N-acetyl-D-glucosamine residues in a peptidoglycan and between N-acetyl-D-glucosamine residues in chitodextrins.
Specifications

Specifications

SPACA5
Polyclonal
PBS with 0.09% sodium azide
Q96QH8
Sperm acrosome-associated protein 5, Lysozyme-like protein 5, Sperm-specific lysozyme-like protein X, SLLP-X, SPACA5, LYZL5, SPACA5A
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
389852
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SPACA5
Liquid
SPACA5
KLH conjugated synthetic peptide between 17-45 amino acids from the N-terminal region of human SPACA5B.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit