Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Space Saver Storage Racks

Economical and convenient storage for samples in screw cap or crimp seal vials. Can be stacked when empty or full, autoclavable at 120°C.

Supplier:  MilliporeSigma™ Supelco™ 23206

 View more versions of this product

Catalog No. 11-102-0835


Explore available promotions
Add to Cart

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Vial Rack
Red
50
Vials (Ø: 29 mm)
336 x 175 x 30 mm
Autoclavable
Polypropylene
Yes
5
30 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

WHEATON is a registered trademark of Wheaton Industries, Inc.