Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPAG1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549729
Description
The antibody detects endogenous level of total SPAG1 protein.
SPAG1 Polyclonal antibody specifically detects SPAG1 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blot.Specifications
SPAG1 | |
Polyclonal | |
Unconjugated | |
SPAG1 | |
CILD28; CT140; epididymis secretory protein Li 268; HEL-S-268; HSD-3.8; Infertility-related sperm protein Spag-1; SP75; SPAG1; sperm associated antigen 1; sperm-associated antigen 1; sperm-associated antigen 1; LOW QUALITY PROTEIN: sperm-associated antigen 1; tetratricopeptide repeat-containing protein; TPIS; TPR-containing protein involved in spermatogenesis | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6674 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q07617 | |
SPAG1 | |
Synthetic peptide corresponding to a region derived from 760-774 amino acids of Human sperm associated antigen 1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction