Learn More
SPAG4 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA549730
Catalog No. PA549730
Add to cart
Description
Description
The antibody detects endogenous level of total SPAG4 protein.
The mammalian sperm flagellum contains two cytoskeletal structures associated with the axoneme: the outer dense fibers surrounding the axoneme in the midpiece and principal piece and the fibrous sheath surrounding the outer dense fibers in the principal piece of the tail. Defects in these structures are associated with abnormal tail morphology, reduced sperm motility, and infertility. In the rat, the protein encoded by this gene associates with an outer dense fiber protein via a leucine zipper motif and localizes to the microtubules of the manchette and axoneme during sperm tail development.
Specifications
Specifications
SPAG4
Polyclonal
Unconjugated
SPAG4
Liquid
SPAG4
Synthetic peptide corresponding to a region derived from 422-436 amino acids of Human sperm associated antigen 4
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
ELISA, Immunohistochemistry (Paraffin), Western Blot
1.7 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q9JJF2, Q9NPE6
SUN4
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
245865, 6676