Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spaglumic acid, Tocris Bioscience™

Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
3106-85-2
C11H14MgN2O8
326.54
MFCD00076652
BQAXDBLSAIPBND-LEUCUCNGSA-L
isospaglumic acid, spaglumic acid, naag, n-acetylaspartylglutamate, n-acetyl-asp-glu, unii-1w8m12wxyl, n-acetylaspartylglutamic acid, 1w8m12wxyl, n-acetyl-l-aspartyl-l-glutamic acid, n-acetyl-l-alpha-aspartyl-l-glutamic acid
188803
CHEBI:73688
magnesium(2+) (4S)-4-carboxy-4-[(2S)-3-carboxylato-2-acetamidopropanamido]butanoate
[Mg++].CC(=O)N[C@@H](CC([O-])=O)C(=O)N[C@@H](CCC([O-])=O)C(O)=O
This item is not returnable. View return policy
3106-85-2
C11H14MgN2O8
326.54
MFCD00076652
BQAXDBLSAIPBND-LEUCUCNGSA-L
isospaglumic acid, spaglumic acid, naag, n-acetylaspartylglutamate, n-acetyl-asp-glu, unii-1w8m12wxyl, n-acetylaspartylglutamic acid, 1w8m12wxyl, n-acetyl-l-aspartyl-l-glutamic acid, n-acetyl-l-alpha-aspartyl-l-glutamic acid
188803
CHEBI:73688
magnesium(2+) (4S)-4-carboxy-4-[(2S)-3-carboxylato-2-acetamidopropanamido]butanoate
[Mg++].CC(=O)N[C@@H](CC([O-])=O)C(=O)N[C@@H](CCC([O-])=O)C(O)=O

Selective mGlu3 agonist

The most abundant peptide neurotransmitter in the mammalian CNS. A weak activator of NMDA receptors, and a highly selective agonist for mGlu3 receptors. Neuroprotective under non-hydrolysing conditions in vivo.
Formula Weight 304.26
Percent Purity >96%
Quantity 50 mg
Chemical Name or Material Spaglumic acid

Recommended Storage : Store at -20°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit