Learn More
SPANXB1 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 170721AP
Catalog No. 50-173-1367
Add to cart
Description
Description
Temporally regulated transcription and translation of several testis-specific genes is required to initiate the series of molecular and morphological changes in the male germ cell lineage necessary for the formation of mature spermatozoa. This gene is a member of the SPANX family of cancer/testis-associated genes, which are located in a cluster on chromosome X. The SPANX genes encode differentially expressed testis-specific proteins that localize to various subcellular compartments. This particular gene maps to chromosome X in a head-to-tail orientation with SPANX family member B2, which appears to be a duplication of the B1 locus. The SPANXB genes are unique members of this gene family, since they contain an additional 18 nt in their coding region compared to the majority of family members. Although the protein encoded by this gene contains consensus nuclear localization signals, the major site for subcellular localization of expressed protein is in the cytoplasmic droplets of ejaculated spermatozoa. This protein provides a biochemical marker for studying the unique structures in spermatazoa, while attempting to further define its role in spermatogenesis.
Specifications
Specifications
SPANXB1
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9NS25
B1, Cancer/testis antigen 11.2, CT11.2, SPANX B, SPANX F, SPANX family member B/F, SPANX family, member B1, SPANXB, SPANXB1, SPANXB2, SPANXF1
Rabbit
IgG
150 μL
-20°C
Primary
728695
Western Blot
Unconjugated
SPANXB1
Liquid
SPANXB1
SPANXB1 Fusion Protein Ag10905
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human