Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPANXC Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP1779250.1MG
Description
SPANXC Polyclonal specifically detects SPANXC in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SPANXC | |
Polyclonal | |
Unconjugated | |
0.02M Potassium Phosphate, 0.15M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
B1, C, cancer/testis antigen 11.3, cancer/testis antigen family 11, member 3, cancer/testis-associated protein CTp11, cancer/testis-associated protein of 11 kD, cancer-testis-associated protein CTp11, CT11.1, CT11.2, CT11.3, CT11.4, CTp11, dJ171K16.1, NAP-X, nuclear-associated protein SPAN-Xc, SPANX, SPANX family, member C, SPAN-Xa, SPANXA, SPANX-A, SPAN-Xb, SPANXB, SPANX-B, SPANX-C, SPAN-Xc protein, SPANX-D, sperm protein associated with the nucleus on the X chromosome C, sperm protein associated with the nucleus, X chromosome, family member C | |
Rabbit | |
Protein A purified | |
Cancer | |
64663 | |
Reconstitute with 100μL deionized water. | |
Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
LYOPH | |
Western Blot 1:1000-1:5000, ELISA 1:5000-1:20000, Immunohistochemistry 1.25-2.5μg/ml, Immunohistochemistry-Paraffin 1:10-1:500 | |
Q9NY87 | |
SPANXC | |
This Protein A purified antibody was prepared from whole rabbit serum produced by repeated immunizations with full-length recombinant human SPANXC protein. | |
0.1 mg | |
Primary | |
This Protein A purified antibody is directed against human SPANX-C protein. | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction